N-Phenyl-m-toluidine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methyldiphenylamine
075291-10G
075291-100G
102540-692EA
64.62
USD
102540-692
102540-694
N-Phenyl-m-toluidine ≥95%
N-Phenyl-m-toluidine
Hazard: Irritant
Formula:
C₁₃H₁₃N MW: 183.25 g/mol Boiling Pt: 317 °C Melting Pt: 25…27 °C Density: 1.066 Flash Pt: 147 °C |
MDL Number:
MFCD00008530 CAS Number: 1205-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...