5-Amino-2-methylphenol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-o-cresol, 3-Hydroxy-4-methylaniline, 1-Amino-3-hydroxy-4-methylbenzene, 5-Amino-2-methylphenol, 4-Amino-2-hydroxytoluene

075289-10G 075289-100G
102540-686EA 33.85 USD
102540-686 102540-688
5-Amino-2-methylphenol ≥95%
5-Amino-2-methylphenol

Hazard: Irritant

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259
Melting Pt: 162
MDL Number: MFCD00043922
CAS Number: 2835-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR