1,3-Dimethoxypropan-2-ol ≥95%

Supplier: Matrix Scientific

075288-1G 075288-5G 075288-10G
102540-680EA 193.85 USD
102540-680 102540-682 102540-684
1,3-Dimethoxypropan-2-ol ≥95%
1,3-Dimethoxypropan-2-ol

Hazard: Irritant

Formula: C₅H₁₂O₃
MW: 120.1500 g/mol
MDL Number: MFCD01076303
CAS Number: 623-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR