4'-Amino-3',5'-dichloroacetophenone ≥95%
Supplier: Matrix Scientific
075287-1G
075287-5G
075287-10G
102540-674EA
55.38
USD
102540-674
102540-676
102540-678
4'-Amino-3',5'-dichloroacetophenone ≥95%
4'-Amino-3',5'-dichloroacetophenone
Hazard: Irritant
Formula:
C₈H₇Cl₂NO |
MDL Number:
MFCD00238535 CAS Number: 37148-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...