(S)-(+)-Methyl mandelate ≥95%
Supplier: Matrix Scientific
Synonyms:
L(+)-Mandelic acid methyl ester, L(+)-2-Hydroxy-2-phenylacetic acid methyl ester, Methyl (S)-(+)-mandelate, L(+)-Methyl mandelate, (S)-(+)-Mandelic acid methyl ester, (S)-(+)-2-Hydroxy-2-phenylacetic acid methyl ester, L(+)-Methyl glycolate, (S)-(+)-Methyl glycolate
075284-10G
075284-100G
102540-662EA
161.54
USD
102540-662
102540-664
(S)-(+)-Methyl mandelate ≥95%
(S)-(+)-Methyl mandelate
Hazard: Irritant
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 58 |
MDL Number:
MFCD00064246 CAS Number: 21210-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...