Methyl-1,2,3,4-tetrahydroisoquinoline-7-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1,2,3,4-tetrahydroisoquinoline-7-carboxylate hydrochloride

075283-250MG 075283-5G
102540-656EA 300 USD
102540-656 102540-660
Methyl-1,2,3,4-tetrahydroisoquinoline-7-carboxylate hydrochloride ≥95%
Methyl-1,2,3,4-tetrahydroisoquinoline-7-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₂
MW: 227.6900 g/mol
MDL Number: MFCD03094735
CAS Number: 220247-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR