Methyl-DL-pyroglutamate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl DL-pyroglutamate, 5-Oxo-pyrrolidine-2-carboxylic acid methyl ester, Methyl 5-oxopyrrolidine-2-carboxylate, Methyl 2-pyrrolidinone-5-carboxylate
075282-5G
075282-25G
102540-652EA
33.85
USD
102540-652
102833-800
Methyl-DL-pyroglutamate ≥95%
Methyl-DL-pyroglutamate
Hazard: Irritant
Formula:
C₆H₉NO₃ MW: 143.14 g/mol |
MDL Number:
MFCD03844651 CAS Number: 54571-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...