2-(3-Chlorophenyl)ethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chlorophenethyl alcohol
075279-1G
075279-5G
102540-634EA
61.54
USD
102540-634
102540-636
2-(3-Chlorophenyl)ethanol ≥95%
2-(3-Chlorophenyl)ethanol
Hazard: Irritant
Formula:
C₈H₉ClO MW: 156.61 g/mol |
MDL Number:
MFCD00002892 CAS Number: 5182-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...