2,4-Dimethylquinazoline ≥95%

Supplier: Matrix Scientific

075276-250MG 075276-1G
102540-620EA 826.15 USD
102540-620 102540-622
2,4-Dimethylquinazoline ≥95%
2,4-Dimethylquinazoline

Hazard: Irritant

Formula: C₁₀H₁₀N₂
MW: 158.2000 g/mol
MDL Number: MFCD18449069
CAS Number: 703-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR