2-Chloro-5-formyl-4-picoline ≥95%

Supplier: Matrix Scientific

075274-250MG 075274-1G 075274-5G
102540-610EA 104.62 USD
102540-610 102540-612 102540-614
2-Chloro-5-formyl-4-picoline ≥95%
2-Chloro-5-formyl-4-picoline

Hazard: Irritant

Formula: C₇H₆ClNO
MW: 155.58 g/mol
MDL Number: MFCD07368881
CAS Number: 884495-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR