3-(2-Aminoethyl)-N-methyl-1H-indole-5-methanesulfonamide ≥95%

Supplier: Matrix Scientific

075270-1G 075270-5G
102540-594EA 533.85 USD
102540-594 102540-596
3-(2-Aminoethyl)-N-methyl-1H-indole-5-methanesulfonamide ≥95%
3-(2-Aminoethyl)-N-methyl-1H-indole-5-methanesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂S
CAS Number: 88919-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR