4-Fluoro-N-methylbenzylamine ≥95%

Supplier: Matrix Scientific

075267-1G 075267-5G 075267-10G
102540-578EA 72.31 USD
102540-578 102540-580 102540-582
4-Fluoro-N-methylbenzylamine ≥95%
4-Fluoro-N-methylbenzylamine

Hazard: Irritant/Air Sensitive

Formula: C₈H₁₀FN
Boiling Pt: 180…181 °C
Density: 1.056
Flash Pt: 71 °C
MDL Number: MFCD03839848
CAS Number: 405-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR