(S)-1-Methylpyrrolidine-2-carboxylic acid hydrate ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Me-Pro-OH·H₂O, N-Methyl-L-proline Monohydrate
075265-1G
075265-5G
075265-10G
102540-568EA
69.23
USD
102540-568
102540-570
102540-572
(S)-1-Methylpyrrolidine-2-carboxylic acid hydrate ≥95%
(S)-1-Methylpyrrolidine-2-carboxylic acid hydrate
Hazard: Irritant
Formula:
C₆H₁₃NO₃ MW: 147.1700 g/mol |
MDL Number:
MFCD00149962 CAS Number: 199917-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...