(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene, (S)-3-(Methylamino)-1-(2-thienyl)-1-propanol

075264-1G 075264-5G
102540-562EA 33.85 USD
102540-562 102540-564
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol ≥95%
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol

Hazard: Irritant

Formula: C₈H₁₃NOS
MW: 171.2600 g/mol
MDL Number: MFCD07357308
CAS Number: 116539-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR