3-(4-Hydroxyphenyl)propionic acid ≥95%

Supplier: Matrix Scientific

075263-100G 075263-500G
102540-560EA 93.85 USD
102540-560 102833-798
3-(4-Hydroxyphenyl)propionic acid ≥95%
3-(4-Hydroxyphenyl)propionic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
MDL Number: MFCD00002778
CAS Number: 501-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR