2-(Methylamino)acetonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

075257-10G 075257-100G
102540-530EA 15.38 USD
102540-530 102540-532
2-(Methylamino)acetonitrile hydrochloride ≥95%
2-(Methylamino)acetonitrile hydrochloride

Hazard: Irritant

Formula: C₃H₇ClN₂
MW: 106.55 g/mol
MDL Number: MFCD00012587
CAS Number: 25808-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR