(±)-2-Chloromandelic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloromandelic acid, (±)-2-Chlorophenylglycolic acid

075256-10G 075256-100G 075256-500G
102540-524EA 15.38 USD
102540-524 102540-526 102540-528
(±)-2-Chloromandelic acid ≥95%
(±)-2-Chloromandelic acid

Hazard: Irritant

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD00084962
CAS Number: 10421-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR