2,6-Dihydroxybenzoic acid ≥95%

Supplier: Matrix Scientific

075254-100G 075254-500G
102540-514EA 60 USD
102540-514 102540-516
2,6-Dihydroxybenzoic acid ≥95%
2,6-Dihydroxybenzoic acid

Hazard: Irritant

Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: ca 164°Cdec
MDL Number: MFCD00002462
CAS Number: 303-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR