2-Methyl-5-nitroanisole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methoxy-2-methyl-5-nitrobenzene, Methyl 5-nitro-o-tolyl ether, 2-Methoxy-4-nitrotoluene

075253-10G 075253-100G
102540-508EA 44.62 USD
102540-508 102540-510
2-Methyl-5-nitroanisole ≥95%
2-Methyl-5-nitroanisole

Hazard: Irritant

Formula: C₈H₉NO₃
MDL Number: MFCD00043912
CAS Number: 13120-77-9
UN: 2446
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR