N-Acetyl-L-methionine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH

075252-10G 075252-100G
102540-504EA 90.77 USD
102540-504 102540-506
N-Acetyl-L-methionine ≥95%
N-Acetyl-L-methionine

Hazard: Irritant

Formula: C₇H₁₃NO₃S
MW: 191.2 g/mol
Melting Pt: 103…107 °C
MDL Number: MFCD00064441
CAS Number: 65-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR