N-Carbobenzoxy-DL-alanine ≥95%
Supplier: Matrix Scientific
Synonyms:
Cbz-DL-Ala-OH, N-Cbz-DL-alanine, 2-(((Benzyloxy)carbonyl)amino)propanoic acid, Z-DL-Ala-OH
075251-10G
075251-100G
075251-500G
102540-498EA
90.77
USD
102540-498
102540-500
102540-502
N-Carbobenzoxy-DL-alanine ≥95%
N-Carbobenzoxy-DL-alanine
Hazard: Irritant
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol |
MDL Number:
MFCD00063125 CAS Number: 4132-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...