N-Carbobenzyloxy-D-alanine ≥95%

Supplier: Matrix Scientific

Synonyms: Z-D-Ala-OH, N-Cbz-D-alanine

075250-10G 075250-100G
102540-494EA 193.85 USD
102540-494 102540-496
N-Carbobenzyloxy-D-alanine ≥95%
N-Carbobenzyloxy-D-alanine

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MW: 223.2 g/mol
MDL Number: MFCD00063126
CAS Number: 26607-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR