2,3,4-Trimethoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

075247-10G 075247-100G
102540-482EA 64.62 USD
102540-482 102540-484
2,3,4-Trimethoxybenzaldehyde ≥95%
2,3,4-Trimethoxybenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00003310
CAS Number: 2103-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR