Methyl hydrogen adipate ≥95%

Supplier: Matrix Scientific

Synonyms: 1,6-Hexanedioic acid methyl ester, Adipic acid methyl ester, Adipic acid monomethyl ester, 1,6-Hexanedioic acid monomethyl ester, Monomethyl adipate

075245-10G 075245-100G
102540-472EA 107.69 USD
102540-472 102540-474
Methyl hydrogen adipate ≥95%
Methyl hydrogen adipate

Hazard: Irritant

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
MDL Number: MFCD00004418
CAS Number: 627-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR