2'-Chloroacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Chlorophenyl)ethanone, o-Chloroacetophenone

075239-10G 075239-100G
102540-452EA 36.92 USD
102540-452 102540-454
2'-Chloroacetophenone ≥95%
2'-Chloroacetophenone

Hazard: Irritant

Formula: C₈H₇ClO
MW: 154.6 g/mol
MDL Number: MFCD00000560
CAS Number: 2142-68-9
UN: 3416
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR