3-(3-Nitrophenoxy)propionic acid ≥95%

Supplier: Matrix Scientific

075238-1G 075238-5G 075238-10G
102540-446EA 427.69 USD
102540-446 102540-448 102540-450
3-(3-Nitrophenoxy)propionic acid ≥95%
3-(3-Nitrophenoxy)propionic acid

Hazard: Irritant

Formula: C₉H₉NO₅
MDL Number: MFCD09049926
CAS Number: 91004-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR