Boc-L-isoleucine ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Boc-L-isoleucine, Boc-Ile-OH
075234-10G
075234-100G
102540-434EA
55.38
USD
102540-434
102540-436
Boc-L-isoleucine ≥95%
Boc-L-isoleucine
Hazard: Irritant
Formula:
C₁₁H₂₁NO₄ |
MDL Number:
MFCD00038324 CAS Number: 13139-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...