4-Iodo-3-methylphenol ≥95%

Supplier: Matrix Scientific

075231-250MG 075231-1G 075231-5G
102540-418EA 203.08 USD
102540-418 102540-420 102540-422
4-Iodo-3-methylphenol ≥95%
4-Iodo-3-methylphenol

Hazard: Irritant

Formula: C₇H₇IO
MW: 234.0300 g/mol
MDL Number: MFCD09029708
CAS Number: 133921-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR