Methyl-2,5-dihydroxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Gentisic acid methyl ester, Methyl 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid methyl ester

075229-5G 075229-10G 075229-100G
102540-406EA 126.15 USD
102540-406 102540-408 102540-410
Methyl-2,5-dihydroxybenzoate ≥95%
Methyl-2,5-dihydroxybenzoate

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00016464
CAS Number: 2150-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR