p-Tolylacetonitrile ≥95%

Supplier: Matrix Scientific

075228-10G 075228-100G
102540-402EA 64.62 USD
102540-402 102540-404
p-Tolylacetonitrile ≥95%
p-Tolylacetonitrile

Hazard: Toxic

Formula: C₉H₉N
Boiling Pt: 242…243 °C
Melting Pt: 18 °C
Density: 0.989
Flash Pt: 106 °C
MDL Number: MFCD00001922
CAS Number: 2947-61-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR