Methyl picolinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl pyridine-2-carboxylate
075225-10G
075225-100G
102540-388EA
38.46
USD
102540-388
102540-390
Methyl picolinate ≥95%
Methyl picolinate
TCA listed: T
Hazard: Irritant
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 95 °C (1 mmHg) Melting Pt: 19 °C Density: 1.137 Flash Pt: 103 °C |
MDL Number:
MFCD00038038 CAS Number: 2459-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...