Ethyl-3-iodopropanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-iodopropanoate

075222-1G 075222-5G
102540-374EA 133.85 USD
102540-374 102540-376
Ethyl-3-iodopropanoate ≥95%
Ethyl-3-iodopropanoate

Hazard: Irritant

Formula: C₅H₉IO₂
MDL Number: MFCD00045323
CAS Number: 6414-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR