2,5-Dibromobenzothiazole ≥95%

Supplier: Matrix Scientific

075221-250MG 075221-1G 075221-5G
102540-368EA 456.92 USD
102540-368 102540-370 102540-372
2,5-Dibromobenzothiazole ≥95%
2,5-Dibromobenzothiazole

Hazard: Irritant

Formula: C₇H₃Br₂NS
MDL Number: MFCD09749228
CAS Number: 1019111-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR