Hydroxyterephthalic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxyterephthalic acid

075220-1G 075220-5G
102540-364EA 129.23 USD
102540-364 102540-366
Hydroxyterephthalic acid ≥95%
Hydroxyterephthalic acid

Hazard: Irritant

Formula: C₈H₆O₅
MW: 182.1302 g/mol
Melting Pt: 311-312°C
MDL Number: MFCD09835368
CAS Number: 636-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR