Quinazoline ≥95%

Supplier: Matrix Scientific

075219-1G 075219-5G 075219-10G
102540-356EA 90.77 USD
102540-356 102540-358 102540-360
Quinazoline ≥95%
Quinazoline

Hazard: Irritant

Formula: C₈H₆N₂
MW: 130.15 g/mol
MDL Number: MFCD00006712
CAS Number: 253-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR