trans-4-Cyclohexyl-L-proline ≥95%

Supplier: Matrix Scientific

Synonyms: (2S,4S)-4-Cyclohexylpyrrolidine-2-carboxylic acid

075217-1G 075217-5G
102540-346EA 229.23 USD
102540-346 102540-348
trans-4-Cyclohexyl-L-proline ≥95%
trans-4-Cyclohexyl-L-proline

Hazard: Irritant

Formula: C₁₁H₁₉NO₂
MW: 197.27 g/mol
MDL Number: MFCD08061929
CAS Number: 103201-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR