2-Acetylphenanthrene ≥95%

Supplier: Matrix Scientific

075216-1G 075216-5G
102540-342EA 200 USD
102540-342 102540-344
2-Acetylphenanthrene ≥95%
2-Acetylphenanthrene

Hazard: Irritant

Formula: C₁₆H₁₂O
MW: 220.2700 g/mol
MDL Number: MFCD00001171
CAS Number: 5960-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR