2-Fluoro-4-morpholinobenzaldehyde ≥95%

Supplier: Matrix Scientific

075210-1G 075210-5G
102540-312EA 640 USD
102540-312 102540-314
2-Fluoro-4-morpholinobenzaldehyde ≥95%
2-Fluoro-4-morpholinobenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₂FNO₂
MW: 209.2169 g/mol
MDL Number: MFCD06656556
CAS Number: 554448-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR