2-Bromo-6-(4-morpholino)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-6-morpholinopyridine

075209-1G 075209-5G 075209-10G
102540-306EA 143.08 USD
102540-306 102540-308 102540-310
2-Bromo-6-(4-morpholino)pyridine ≥95%
2-Bromo-6-(4-morpholino)pyridine

Hazard: Irritant

Formula: C₉H₁₁BrN₂O
MW: 243.1004 g/mol
MDL Number: MFCD07772824
CAS Number: 332134-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR