DL-α-Phenylglycine ≥95%

Supplier: Matrix Scientific

Synonyms: Dl-A-Aminophenylacetic Acid, H-DL-Phg-OH, DL-2-Phenylglycine

075208-100G 075208-500G
102540-302EA 86.15 USD
102540-302 102540-304
DL-α-Phenylglycine ≥95%
DL-α-Phenylglycine

TCA listed: T
Hazard: Irritant

Formula: C₈H₉NO₂
Melting Pt: 290°C
MDL Number: MFCD00064402
CAS Number: 2835-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR