(R)-tert-Butyl-2-carbamoylpyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-tert-Butyl 2-carbamoylpyrrolidine-1-carboxylate, 1-Boc-D-prolinamide

075206-1G 075206-5G 075206-10G
102540-292EA 21.54 USD
102540-292 102540-294 102540-296
(R)-tert-Butyl-2-carbamoylpyrrolidine-1-carboxylate ≥95%
(R)-tert-Butyl-2-carbamoylpyrrolidine-1-carboxylate

Purity: 95+%
CAS: 70138-72-6
MDL Number: MFCD03419274
Molecular Formula: C10H18N2O3
Molecular Weight: 214,27
Hazard: Irritant

Formula: C₁₀H₁₈N₂O₃
MW: 214.27 g/mol
Melting Pt: 101…102 °C
MDL Number: MFCD03419274
CAS Number: 70138-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR