4,6-Dichloro-2-(trifluoromethyl)quinazoline ≥95%

Supplier: Matrix Scientific

075204-250MG 075204-1G 075204-5G
102540-282EA 456.92 USD
102540-282 102540-284 102540-286
4,6-Dichloro-2-(trifluoromethyl)quinazoline ≥95%
4,6-Dichloro-2-(trifluoromethyl)quinazoline

Hazard: Irritant

Formula: C₉H₃Cl₂F₃N₂
CAS Number: 746671-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR