2,4-Dichlorobenzoic acid ≥95%
Supplier: Matrix Scientific
075202-10G
075202-100G
102540-274EA
33.85
USD
102540-274
102540-276
2,4-Dichlorobenzoic acid ≥95%
2,4-Dichlorobenzoic acid
Hazard: Irritant
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol |
MDL Number:
MFCD00002414 CAS Number: 50-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...