Sodium-2-(2-amino-3-benzoylphenyl)acetate ≥95%

Supplier: Matrix Scientific

075197-1G 075197-5G
102540-256EA 213.85 USD
102540-256 102540-258
Sodium-2-(2-amino-3-benzoylphenyl)acetate ≥95%
Sodium-2-(2-amino-3-benzoylphenyl)acetate

Hazard: Irritant

Formula: C₁₅H₁₂NNaO₃
MW: 277.26 g/mol
MDL Number: MFCD09842317
CAS Number: 61941-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR