Cladribine ≥95%

Supplier: Matrix Scientific

Synonyms: (2R,3S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol, CdA, 2-Chloro-2'-deoxyadenosine, NSC105014, RWJ 26251

075196-250MG 075196-1G
102540-252EA 1066.15 USD
102540-252 102540-254
Cladribine ≥95%
Cladribine

Hazard: Irritant

Formula: C₁₀H₁₂ClN₅O₃
MW: 285.7 g/mol
MDL Number: MFCD00153939
CAS Number: 4291-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR