5-Bromo-3-fluoro-4-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

075194-1G 075194-5G
102540-242EA 230.77 USD
102540-242 102540-244
5-Bromo-3-fluoro-4-methylbenzaldehyde ≥95%
5-Bromo-3-fluoro-4-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₆BrFO
MDL Number: MFCD18205969
CAS Number: 1370411-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR