(4-(3-Methoxypropoxy)-3-methylpyridin-2-yl)methanol ≥95%

Supplier: Matrix Scientific

075193-250MG 075193-1G 075193-5G
102540-236EA 355.38 USD
102540-236 102540-238 102540-240
(4-(3-Methoxypropoxy)-3-methylpyridin-2-yl)methanol ≥95%
(4-(3-Methoxypropoxy)-3-methylpyridin-2-yl)methanol

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD08274729
CAS Number: 118175-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR