6-Fluoro-4-hydroxyquinoline-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

075190-1G 075190-5G
102540-222EA 213.85 USD
102540-222 102540-224
6-Fluoro-4-hydroxyquinoline-3-carboxylic acid ≥95%
6-Fluoro-4-hydroxyquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆FNO₃
MW: 207.1600 g/mol
MDL Number: MFCD00973836
CAS Number: 343-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR