5-(Trifluoromethyl)-1H-indazole ≥95%

Supplier: Matrix Scientific

075189-1G 075189-5G
102540-218EA 355.38 USD
102540-218 102540-220
5-(Trifluoromethyl)-1H-indazole ≥95%
5-(Trifluoromethyl)-1H-indazole

Hazard: Irritant

Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
MDL Number: MFCD08234904
CAS Number: 885271-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR