Ethyl-2-oxohexanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-oxohexanoate

075187-1G 075187-5G
102540-208EA 323.08 USD
102540-208 102540-210
Ethyl-2-oxohexanoate ≥95%
Ethyl-2-oxohexanoate

Hazard: Irritant

Formula: C₈H₁₄O₃
MDL Number: MFCD09952021
CAS Number: 5753-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR